Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

642002

Sigma-Aldrich

(3S,5R,8aS)-(+)-Hexahydro-3-phenyl-5H-oxazolo[3,2-a]pyridine-5-carbonitrile

97%

Synonym(s):

(+)-2-Cyano-6-phenyloxazolopiperidine, [3S-(3α,5β,8aβ)]-Hexahydro-3-phenyl-5H-oxazolo[3,2-a]pyridine-5-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

79-83 °C (lit.)

SMILES string

[H][C@]12CCC[C@H](C#N)N1[C@H](CO2)c3ccccc3

InChI

1S/C14H16N2O/c15-9-12-7-4-8-14-16(12)13(10-17-14)11-5-2-1-3-6-11/h1-3,5-6,12-14H,4,7-8,10H2/t12-,13-,14+/m1/s1

InChI key

GQHMNZGZXHZLEN-MCIONIFRSA-N

Application

(3S,5R,8aS)-(+)-Hexahydro-3-phenyl-5H-oxazolo[3,2-a]pyridine-5-carbonitrile can be used as a starting material:
  • To prepare spirocyclic aminochroman derivatives as possible 5-HT1A agonists.
  • In the asymmetric synthesis of aza-analogs of podophyllotoxin as potent antitumor agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of spirocyclic aminochroman derivatives according to the CN (R, S) strategy
Pave G, et al.
The Journal of Organic Chemistry, 68(4), 1401-1408 (2003)
Asymmetric synthesis XXVIII: Hydroxylated benzoquinolizidine analogues of podophyllotoxin via the (CN (R, S) method.
Lienard P, et al.
Tetrahedron, 49(19), 3995-4006 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service