Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

641103

Sigma-Aldrich

2-(Triisopropylsilyl)oxazole-5-carboxaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23NO2Si
CAS Number:
Molecular Weight:
253.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

refractive index

n20/D 1.5030 (lit.)

density

0.988 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cnc(o1)[Si](C(C)C)(C(C)C)C(C)C

InChI

1S/C13H23NO2Si/c1-9(2)17(10(3)4,11(5)6)13-14-7-12(8-15)16-13/h7-11H,1-6H3

InChI key

FBQMBIDSILZYSW-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service