Skip to Content
MilliporeSigma
All Photos(1)

Documents

640549

Sigma-Aldrich

3-Methoxytriphenylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO
CAS Number:
Molecular Weight:
275.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.6550 (lit.)

bp

130-135 °C/0.6 mmHg (lit.)

SMILES string

COc1cccc(c1)N(c2ccccc2)c3ccccc3

InChI

1S/C19H17NO/c1-21-19-14-8-13-18(15-19)20(16-9-4-2-5-10-16)17-11-6-3-7-12-17/h2-15H,1H3

InChI key

MVIPGWFTBPVNBJ-UHFFFAOYSA-N

Application

3-Methoxy triphenylamine can be used in the synthesis of Schiff base molecules, which can be used as fluorescence and colorimetric chemosensors to selectively detect zinc(II) and cadmium(II) ions in acetonitrile/aqueous solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Selective turn-on fluorescence for Zn 2+ and Zn 2++ Cd 2+ metal ions by single Schiff base chemosensor"
Hariharan.S.P and Anthony PS
Analytica Chimica Acta, 848, 74-79 (2014)
Anu Kundu et al.
Journal of fluorescence, 29(6), 1359-1369 (2019-11-16)
New series of methoxy and hydroxyl group substituted triphenylamine (TPA)-imidazole fluorescent molecules (5-(diphenylamino)-2-(1H-phenanthro[9,10-d]imidazol-2-yl)phenol (1), 5-(diphenylamino)-2-(1-phenyl-1H-phenanthro[9,10-d]imidazol-2-yl)phenol (2), 5-(diphenylamino)-2-(4,5-diphenyl-1H-imidazol-2-yl)phenol (3), 5-(diphenylamino)-2-(1,4,5-triphenyl-1H-imidazol-2-yl)phenol (4), N-(3-methoxy-4-(1H-phenanthro[9,10-d]imidazol-2-yl)phenyl)-N-phenylbenzenamine (5), N-(3-methoxy-4-(1-phenyl-1H-phenanthro[9,10-d]imidazol-2-yl)phenyl)-N-phenylbenzene amine (6), and N-(3-methoxy-4-(4,5-diphenyl-1H-imidazol-2-yl)phenyl)-N-phenylbenzenamine (7)) have been synthesized that exhibited strong solution fluorescence and molecular structure and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service