Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

639362

Sigma-Aldrich

2-(Ethylhexyl)lithium solution

30-35 wt. % in heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCH2CH(CH2CH3)(CH2)3CH3
CAS Number:
Molecular Weight:
120.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

30-35 wt. % in heptane

density

0.730 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CC(CC)CCCC

InChI

1S/C8H17.Li/c1-4-6-7-8(3)5-2;/h8H,3-7H2,1-2H3;

InChI key

VKMNMYVWVWHTLX-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service