Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

639125

Sigma-Aldrich

2-Biphenylmagnesium bromide solution

0.5 M in diethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$361.00

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$361.00

About This Item

Linear Formula:
C6H5C6H4MgBr
CAS Number:
Molecular Weight:
257.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

0.5 M in diethyl ether

density

0.800 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

Br[Mg]c1ccccc1-c2ccccc2

InChI

1S/C12H9.BrH.Mg/c1-3-7-11(8-4-1)12-9-5-2-6-10-12;;/h1-9H;1H;/q;;+1/p-1

InChI key

NCZXESZVEDNTHU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service