Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

639028

Sigma-Aldrich

4-Bromo-5-fluoro-2-iodotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrFI
CAS Number:
Molecular Weight:
314.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.624 (lit.)

bp

249-250 °C (lit.)

density

2.145 g/mL at 25 °C (lit.)

functional group

bromo
fluoro
iodo

SMILES string

Cc1cc(F)c(Br)cc1I

InChI

1S/C7H5BrFI/c1-4-2-6(9)5(8)3-7(4)10/h2-3H,1H3

InChI key

ACDTXEFSNCPJED-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service