Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

638765

Sigma-Aldrich

3-Chloro-4-methoxycinnamic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClO3
CAS Number:
Molecular Weight:
212.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

SMILES string

COc1ccc(\C=C\C(O)=O)cc1Cl

InChI

1S/C10H9ClO3/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6H,1H3,(H,12,13)/b5-3+

InChI key

FFQJKQJQLDOHDW-HWKANZROSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service