Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63868

Sigma-Aldrich

(Chloromethyl)polystyrene

porous, extent of labeling: ~5.5 mmol/g Cl loading

Synonym(s):

Merrifield resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

porous

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

~5.5 mmol/g Cl loading

matrix

crosslinked with 5.5% DVB

particle size

16-50 mesh

application(s)

peptide synthesis

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service