Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636975

Sigma-Aldrich

2,5-Dioctyl-1,4-di-1-propynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42
CAS Number:
Molecular Weight:
378.63
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

53-57 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCc1cc(C#CC)c(CCCCCCCC)cc1C#CC

InChI

1S/C28H42/c1-5-9-11-13-15-17-21-27-23-26(20-8-4)28(24-25(27)19-7-3)22-18-16-14-12-10-6-2/h23-24H,5-6,9-18,21-22H2,1-4H3

InChI key

SKVLNUXBFHZNAZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service