Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

635111

Sigma-Aldrich

Strontium lanthanum aluminate

single crystal substrate, <110>, ≥99.9% trace metals basis

Synonym(s):

Strontium lanthanum aluminum oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrLaAlO4
CAS Number:
Molecular Weight:
317.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

single side polished

Assay

≥99.9% trace metals basis

dielectric constant

16.8

size

10 mm × 10 mm × 0.5 mm

mp

1650 °C (lit.)

density

5.92 g/mL at 25 °C (lit.)

SMILES string

O1[Sr]O[La]2O[Al]1O2

InChI

1S/Al.La.4O.Sr

InChI key

QOBCMFRTIKRXKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Excellent substrate for high temperature superconductor thin films.

Physical form

tetragonal (a = 3.756 Å, c = 12.63 Å)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service