Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

635103

Sigma-Aldrich

Yttrium vanadate

single crystal substrate, <110>, 99.99% trace metals basis

Synonym(s):

Vanadium yttrium oxide, Yttrium vanadium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YVO4
CAS Number:
Molecular Weight:
203.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

single side polished

Assay

99.99% trace metals basis

hardness

5 mohs

size

10 mm × 10 mm × 0.5 mm

transmittance

400-4000 nm, ~80%

mp

1825 °C (lit.)

density

4.22 g/mL at 25 °C (lit.)

SMILES string

[Y+3].[O-][V]([O-])([O-])=O

Features and Benefits

Birefringent yttrium vanadate crystals are ideal materials for optical polarizing components such as fiber optics or beam displacers. It is transparent over a large range and is a material of medium hardness and can therefore be polished to a high optical finish.

Physical properties

Refractive Index η, Birefringence (Δn) and walk-off angle at 45° (ρ):
η o= 1.9929 η e= 2.2154 Δη = 0.2225 ρ=6.04° at λ 0.63 μm
η o= 1.9500 η e= 2.1554 Δη = 0.2054 ρ=5.72° at λ 1.30 μm
η o= 1.9447 η e= 2.1486 Δη = 0.2039 ρ=5.69° at λ 1.55 μm
Thermal Conductivity: A axis 5.32 W/(m.k), C axis = 5.10 W/(m.k)
Thermal Expansion: αa= 4.43 x10-6/ °C, αc= 11.37 x10-6/ °C
Thermal Optic Coefficient: dη a / dT : 8.5x10-6/ °C, dη c / dT : 3.0 x10-6/ °C

Physical form

tetragonal (a = b = 7.12 Å, c = 6.29 Å)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service