Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

634697

Sigma-Aldrich

Magnesium oxide

(single crystal substrate), <111>, ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

single side polished

Assay

≥99.9% trace metals basis

form

(single crystal substrate)

dielectric constant

9.8

reaction suitability

reagent type: catalyst
core: magnesium

L × W × thickness

10 mm × 10 mm × 0.5 mm

impurities

<10 ppm C
<10 ppm Cr
<10 ppm Si
<15 ppm Al
<40 ppm Ca
<5 ppm B
<50 ppm Fe

transmittance

200-400 nm, >90%
500-1000 nm, >98%

mp

2852 °C (lit.)

semiconductor properties

<111>

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Magnesium oxide is an excellent single-crystal substrate for thin films of ferromagnetic, photo-electronic, and high Tc superconductor materials.

Physical properties

Thermal expansion: 12.8 x 10-6/°C

Physical form

cubic (a = 4.216 Å)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Sun et al.
BMC plant biology, 15, 116-116 (2015-05-11)
Heat stress is a critical threat to tall fescue in transitional and warm climate zones. Identification of association between molecular markers and heat tolerance-related functional traits would promote the efficient selection of heat tolerant tall fescue cultivars. Association analysis of
Benjamin Michen et al.
Environmental science & technology, 47(3), 1526-1533 (2013-01-05)
Ceramic filters, working on the depth filtration principle, are known to improve drinking water quality by removing human pathogenic microorganisms from contaminated water. However, these microfilters show no sufficient barrier for viruses having diameters down to 20 nm. Recently, it
Charles T Campbell et al.
Journal of the American Chemical Society, 134(43), 18109-18115 (2012-10-05)
Adsorbed molecules are involved in many reactions on solid surface that are of great technological importance. As such, there has been tremendous effort worldwide to learn how to predict reaction rates and equilibrium constants for reactions involving adsorbed molecules. Theoretical
Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service