Skip to Content
MilliporeSigma
All Photos(2)

Documents

634476

Sigma-Aldrich

Furan-3-ylethynyltrimethylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12OSi
CAS Number:
Molecular Weight:
164.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4910 (lit.)

bp

184-186 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C#Cc1ccoc1

InChI

1S/C9H12OSi/c1-11(2,3)7-5-9-4-6-10-8-9/h4,6,8H,1-3H3

InChI key

YTKPUDNQUBDSHF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F

Flash Point(C)

65 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service