Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

634255

Sigma-Aldrich

Terbium(III,IV) oxide

nanopowder, <100 nm particle size (BET), 99.5% trace metals basis

Synonym(s):

Terbium peroxide, Tetraterbium heptoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb4O7
CAS Number:
Molecular Weight:
747.70
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: terbium

particle size

<100 nm (BET)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

InChI

1S/7O.4Tb

InChI key

UWZVBPNIJCKCMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service