Skip to Content
MilliporeSigma
All Photos(1)

Documents

633437

Sigma-Aldrich

Diacetonamine hydrogenoxalate

90%

Synonym(s):

4-Amino-4-methyl-2-pentanone hydrogenoxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2C(CH3)2NH2·HO2CCO2H
CAS Number:
Molecular Weight:
115.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

125-130 °C (lit.)

SMILES string

OC(=O)C(O)=O.CC(=O)CC(C)(C)N

InChI

1S/C6H13NO.C2H2O4/c1-5(8)4-6(2,3)7;3-1(4)2(5)6/h4,7H2,1-3H3;(H,3,4)(H,5,6)

InChI key

PABISXFQAHDPMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aki Ikeda et al.
AMB Express, 7(1), 101-101 (2017-05-27)
Under bioassay-guided investigation, a sporulation-inducing factor (SIF) toward Bacillus spp. was searched for in methanol (MeOH) extracts of soybean curd residues, and diacetonamine (1) was identified as the active compound. SIF was first isolated as a monoacetylated derivative (2, 4.1 mg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service