Skip to Content
MilliporeSigma
All Photos(1)

Documents

633259

Sigma-Aldrich

N-Acetoxy-d3-succinimide

98 atom % D

Synonym(s):

Acetic-d3 acid, 2,5-dioxopyrrolidin-1-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D3H4NO4
CAS Number:
Molecular Weight:
160.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

technique(s)

protein expression: suitable

mp

133 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C(=O)ON1C(=O)CCC1=O

InChI

1S/C6H7NO4/c1-4(8)11-7-5(9)2-3-6(7)10/h2-3H2,1H3/i1D3

InChI key

SIFCHNIAAPMMKG-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guijin Zhai et al.
Talanta, 125, 411-417 (2014-05-21)
Mass spectrometric quantification of phosphopeptides is a challenge due to the ion suppression effect of highly abundant non-phosphorylated peptides in complex samples such as serum. Several strategies for relative quantification of serum phosphopeptides based on MS have been developed, but
Yu Lin et al.
Journal of inorganic biochemistry, 146, 44-51 (2015-03-15)
Electrospray ionization mass spectrometry (ESI-MS) has been widely used to identify binding sites of metal complexes to proteins. However, the MS quantification of the metal-protein coordination remains a challenge. We have recently demonstrated by ESI-MS analysis that organometallic ruthenium complexes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service