Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632899

Sigma-Aldrich

2,3-Difluorotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3F2
CAS Number:
Molecular Weight:
128.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4530 (lit.)

bp

119-121 °C (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(F)c1F

InChI

1S/C7H6F2/c1-5-3-2-4-6(8)7(5)9/h2-4H,1H3

InChI key

ZNEHIDGAPGVZSA-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service