Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632031

Sigma-Aldrich

3-(Trimethylsilyloxy)-1-butyne

97%

Synonym(s):

2-(Trimethylsilyloxy)-3-butyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOCH(CH3)C≡CH
CAS Number:
Molecular Weight:
142.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.405 (lit.)

mp

127-131 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(O[Si](C)(C)C)C#C

InChI

1S/C7H14OSi/c1-6-7(2)8-9(3,4)5/h1,7H,2-5H3

InChI key

RMKACVRKUKSWNQ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.9 °F - closed cup

Flash Point(C)

5.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirokazu Muraoka et al.
Cell reports, 27(1), 199-212 (2019-04-04)
Nicotinamide adenine dinucleotide (NAD+) metabolism plays a critical role in kidneys. We previously reported that decreased secretion of a NAD+ precursor, nicotinamide mononucleotide (NMN), from proximal tubules (PTs) can trigger diabetic albuminuria. In the present study, we investigated the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service