Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631671

Sigma-Aldrich

5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-bi-2-naphthol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H10OH)2
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

138-142 °C (lit.)

SMILES string

Oc1ccc2CCCCc2c1-c3c(O)ccc4CCCCc34

InChI

1S/C20H22O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h9-12,21-22H,1-8H2

InChI key

UTXIFKBYNJRJPH-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service