Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631566

Sigma-Aldrich

3-Pyridineboronic acid 1,3-propanediol ester

97%

Synonym(s):

1,3-propanediol cyclic ester, 3-Pyridineboronic acid glycol ester, 3-Pyridyl trimethylene borate, 3-[1,3,2]Dioxaborinan-2-yl-pyridine, Pyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BNO2
CAS Number:
Molecular Weight:
162.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

solid

mp

95-99 °C (lit.)

SMILES string

C1COB(OC1)c2cccnc2

InChI

1S/C8H10BNO2/c1-3-8(7-10-4-1)9-11-5-2-6-12-9/h1,3-4,7H,2,5-6H2

InChI key

ZUUZNOFTKGDLLN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service