Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631302

Sigma-Aldrich

3-(3-Chlorophenyl)propionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(3-Cl-C6H4)CH2CH2CO2H
CAS Number:
Molecular Weight:
184.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

72-76 °C (lit.)

SMILES string

OC(=O)CCc1cccc(Cl)c1

InChI

1S/C9H9ClO2/c10-8-3-1-2-7(6-8)4-5-9(11)12/h1-3,6H,4-5H2,(H,11,12)

InChI key

CLTDVBQNUHHYCA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man Zhang et al.
Heterocycles, 101(1), 145-164 (2020-08-11)
Neuropathic pain, epilepsy, insomnia, and tremor disorder may arrive from an increase of intracellular Ca2+ concentration through a dysfunction of T-type Ca2+ channels. Thus, T-type calcium channels could be a target in drug discovery for the treatments of neuropathic pain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service