Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631221

Sigma-Aldrich

5-(3,4-Dichlorophenyl)-2-furonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H5Cl2NO
CAS Number:
Molecular Weight:
238.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

155-159 °C (lit.)

SMILES string

Clc1ccc(cc1Cl)-c2ccc(o2)C#N

InChI

1S/C11H5Cl2NO/c12-9-3-1-7(5-10(9)13)11-4-2-8(6-14)15-11/h1-5H

InChI key

LLXJJYJKCUDNAV-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service