Skip to Content
MilliporeSigma

630845

Sigma-Aldrich

3-Heptyn-1-ol

98%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

refractive index

n20/D 1.456

bp

80-82 °C/15 mmHg

density

0.920 g/mL at 25 °C

SMILES string

CCCC#CCCO

InChI

1S/C7H12O/c1-2-3-4-5-6-7-8/h8H,2-3,6-7H2,1H3

InChI key

PSWHODJVUOXHKA-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service