Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630802

Sigma-Aldrich

N-Boc-1-aminocyclobutane carboxylic acid

97%

Synonym(s):

1-(Boc-amino)cyclobutanecarboxylic acid, Boc-cyclovaline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHC4H6CO2H
CAS Number:
Molecular Weight:
215.25
Beilstein:
7994003
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

129-133 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NC1(CCC1)C(O)=O

InChI

1S/C10H17NO4/c1-9(2,3)15-8(14)11-10(7(12)13)5-4-6-10/h4-6H2,1-3H3,(H,11,14)(H,12,13)

InChI key

ROVVUKFHORPDSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service