Skip to Content
MilliporeSigma
All Photos(1)

Documents

630403

Sigma-Aldrich

4-Acetoxybutylzinc bromide

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CCO2(CH2)4ZnBr
Molecular Weight:
260.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.985 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OCCCC[Zn]Br

InChI

1S/C6H11O2.BrH.Zn/c1-3-4-5-8-6(2)7;;/h1,3-5H2,2H3;1H;/q;;+1/p-1

InChI key

DBPKJQMTXTUZPP-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service