Skip to Content
MilliporeSigma
All Photos(1)

Documents

630136

Sigma-Aldrich

Poly(2,2,3,3,3-pentafluoropropyl acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5F5O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

refractive index

n20/D 1.3890 (lit.)

transition temp

Tg -26 °C(lit.)

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: soluble

density

1.446 g/mL at 25 °C (lit.)

Application

Low refractive index polymer for cladding layer in optical waveguides. Transparent, low surface tension polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service