Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62633

Sigma-Aldrich

(+)-Longicyclene

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1S,2S)-1,2,6,6-Tetramethyltetracyclo[8.1.0.02.8.07.11]undecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2039581
EC Number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥95.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +32.5±1°, neat

refractive index

n20/D 1.493

bp

252-254 °C (lit.)

density

0.937 g/mL at 20 °C (lit.)

SMILES string

CC1(C)CCC[C@@]2(C)[C@@H]3C[C@@H]4[C@H]([C@H]13)[C@@]24C

InChI

1S/C15H24/c1-13(2)6-5-7-14(3)9-8-10-12(11(9)13)15(10,14)4/h9-12H,5-8H2,1-4H3/t9-,10-,11-,12-,14+,15+/m1/s1

InChI key

WCEIQUQVIOGRBF-YTDSDZJUSA-N

General description

Longicyclene is the first tetracarbocyclic sesquiterpene isolated from the essential oil of Pinus longifolia, Roxb.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies in sesquiterpenes-XXXV: longicyclene, the first tetracyclic sesquiterpene.
Nayak UR and Dev S
Tetrahedron, 24(11), 4099-4104 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service