Skip to Content
MilliporeSigma
All Photos(1)

Documents

61891

Sigma-Aldrich

L-tert-Leucine methyl ester hydrochloride

≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH(NH2)CO2CH3 · HCl
CAS Number:
Molecular Weight:
181.66
Beilstein:
4879785
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-tert-Leucine methyl ester hydrochloride, ≥99.0% (AT)

Assay

≥99.0% (AT)

form

powder or crystals

optical activity

[α]20/D +17.5±1°, c = 1% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

183.0 °C

application(s)

cell analysis
peptide synthesis

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@@H](N)C(C)(C)C

InChI

1S/C7H15NO2.ClH/c1-7(2,3)5(8)6(9)10-4;/h5H,8H2,1-4H3;1H/t5-;/m1./s1

InChI key

HRTQWUHFSXVRPY-NUBCRITNSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service