Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616621

Sigma-Aldrich

2-Keto-4-methyl-d3-pentanoic acid sodium salt

98 atom % D

Synonym(s):

2-Ketoisocaproic acid-d3 (4-methyl-d3) sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$650.00

$650.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 MG
$650.00

About This Item

Linear Formula:
CD3CH(CH3)CH2COCO2Na
Molecular Weight:
155.14
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$650.00


Please contact Customer Service for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

form

solid

availability

not available in Japan

technique(s)

bio NMR: suitable

mp

280 °C (dec.) (lit.)

mass shift

M+3

SMILES string

[Na+].[2H]C([2H])([2H])C(C)CC(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-4(2)3-5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1/i1D3;

InChI key

IXFAZKRLPPMQEO-NIIDSAIPSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service