Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61642

Sigma-Aldrich

Butyl dodecanoate

≥99.0% (GC)

Synonym(s):

Butyl laurate, Lauric acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COO(CH2)3CH3
CAS Number:
Molecular Weight:
256.42
Beilstein:
1776360
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

180 °C/18 mmHg (lit.)

mp

−10 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCCCCCCCC(=O)OCCCC

InChI

1S/C16H32O2/c1-3-5-7-8-9-10-11-12-13-14-16(17)18-15-6-4-2/h3-15H2,1-2H3

InChI key

NDKYEUQMPZIGFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service