Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615234

Sigma-Aldrich

Diphenyl sulfoxide-d10

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6D5)2SO
CAS Number:
Molecular Weight:
212.33
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

bp

206-208 °C/13 mmHg (lit.)

mp

69-71 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])S(=O)c2c([2H])c([2H])c([2H])c([2H])c2[2H]

InChI

1S/C12H10OS/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

JJHHIJFTHRNPIK-LHNTUAQVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service