Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615072

Sigma-Aldrich

2-Propanol-1,1,1-d3

98 atom % D

Synonym(s):

Isopropyl-1,1,1-d3 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH(OH)CH3
CAS Number:
Molecular Weight:
63.11
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

99% (CP)

bp

82 °C

mp

-90 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3/i1D3

InChI key

KFZMGEQAYNKOFK-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service