Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

613835

Sigma-Aldrich

Acetylene-d2

99 atom % D

Synonym(s):

Dideuterioacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2D2
CAS Number:
Molecular Weight:
28.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99 atom % D

mp

-81 °C (subl.) (lit.)

density

0.97 g/mL at 0 °C (air=1)(lit.)

mass shift

M+2

SMILES string

[2H]C#C[2H]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Hazard Classifications

Chem. Unst. Gas A - Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service