Skip to Content
MilliporeSigma
All Photos(1)

Documents

613576

Sigma-Aldrich

Tetramethyl-d12-ammonium chloride

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)4N+Cl-
Molecular Weight:
121.67
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

>300 °C

mass shift

M+12

SMILES string

[2H]C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].[Cl-]

InChI

1S/C4H12N.ClH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1/i1D3,2D3,3D3,4D3;

InChI key

OKIZCWYLBDKLSU-KXWZVCIVSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service