Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

613061

Sigma-Aldrich

Acetic acid-d4

"100%", 99.96 atom % D

Synonym(s):

Acetic-d3 acid-d, Tetradeuteroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CO2D
CAS Number:
Molecular Weight:
64.08
Beilstein:
1748971
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

vapor density

2.07 (vs air)

Quality Level

vapor pressure

11.4 mmHg ( 20 °C)

isotopic purity

99.96 atom % D

Assay

99% (CP)

autoignition temp.

800 °F

expl. lim.

16 %

refractive index

n20/D 1.368 (lit.)

bp

115.5 °C (lit.)

mp

15-16 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]OC(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD

InChI key

QTBSBXVTEAMEQO-GUEYOVJQSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service