Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

612197

Sigma-Aldrich

2,2,2-Trifluoroethanol-1,1-d2

99.5 atom % D

Synonym(s):

2,2,2-Trifluoroethyl-1,1-d2 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CD2OH
CAS Number:
Molecular Weight:
102.05
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

impurities

≤0.05% water
water

bp

77-80 °C (lit.)

mp

-44 °C (lit.)

density

1.373 g/mL

mass shift

M+2

SMILES string

[H]OC([2H])([2H])C(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2/i1D2

InChI key

RHQDFWAXVIIEBN-DICFDUPASA-N

General description

2,2,2-Trifluoroethanol-1,1-d2 (2,2,2-Trifluoroethyl-1,1-d2 alcohol, TFE-d2) is a deutered trifluoroethanol that may be employed as an NMR solvent.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service