Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

609986

Sigma-Aldrich

Boric oxide-18O3

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B218O3
Molecular Weight:
75.62
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

95 atom % 18O

Quality Level

form

solid

mass shift

M+6

SMILES string

[18O]=B[18O]B=[18O]

InChI

1S/B2O3/c3-1-5-2-4/i3+2,4+2,5+2

InChI key

JKWMSGQKBLHBQQ-LXJNWNNVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service