Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

608394

Sigma-Aldrich

Uridine-13C9,15N2 5′-triphosphate sodium salt

90% (CP), 98 atom % 15N, 98 atom % 13C

Synonym(s):

UTP-ul-13C,15N sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H1515N2O15P3
Molecular Weight:
495.06
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

isotopic purity

98 atom % 13C
98 atom % 15N

Assay

90% (CP)

shipped in

dry ice


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service