Skip to Content
MilliporeSigma
All Photos(1)

Documents

607495

Sigma-Aldrich

Formamide-15N,d2

98 atom % 15N, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO15ND2
Molecular Weight:
48.05
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

98 atom % 15N
98 atom % D

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.208 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]N([2H])C=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)/i/hD2

InChI key

ZHNUHDYFZUAESO-ZSJDYOACSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service