Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

606375

Sigma-Aldrich

Iodoform-13C

99 atom % 13C, 99% (CP)

Synonym(s):

Triiodomethane-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CHI3
Molecular Weight:
394.72
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mp

118-121 °C (lit.)

mass shift

M+1

storage temp.

−20°C

SMILES string

I[13CH](I)I

InChI

1S/CHI3/c2-1(3)4/h1H/i1+1

InChI key

OKJPEAGHQZHRQV-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service