Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605905

Sigma-Aldrich

Cholesteryl octanoate-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC34H60O2
Molecular Weight:
513.84
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

110 °C

mass shift

M+1

SMILES string

CCCCCCC[13C](=O)O[C@H]1CC[C@]2(C)C3CC[C@]4(C)[C@H](CCC4C3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C35H60O2/c1-7-8-9-10-11-15-33(36)37-28-20-22-34(5)27(24-28)16-17-29-31-19-18-30(26(4)14-12-13-25(2)3)35(31,6)23-21-32(29)34/h16,25-26,28-32H,7-15,17-24H2,1-6H3/t26-,28+,29+,30-,31+,32+,34+,35-/m1/s1/i33+1

InChI key

SKLBBRQPVZDTNM-SPKDLXGUSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service