Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605700

Sigma-Aldrich

Palmitic acid-5,6,7,8-13C4

99 atom % 13C, 99% (CP)

Synonym(s):

Hexadecanoic-5,6,7,8-13C4 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7(13CH2)4(CH2)3CO2H
Molecular Weight:
260.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

mp

61-64 °C (lit.)

mass shift

M+4

SMILES string

CCCCCCCC[13CH2][13CH2][13CH2][13CH2]CCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i9+1,10+1,11+1,12+1

InChI key

IPCSVZSSVZVIGE-BMHINFNASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service