Skip to Content
MilliporeSigma
All Photos(1)

Documents

604054

Sigma-Aldrich

Dimethyl acetylenedicarboxylate-1,2,3,4-13C4

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CO213C≡13C13CO2CH3
CAS Number:
Molecular Weight:
146.08
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

density

1.156 g/mL at 25 °C

mass shift

M+4

SMILES string

CO[13C](=O)[13C]#[13C][13C](=O)OC

InChI

1S/C6H6O4/c1-9-5(7)3-4-6(8)10-2/h1-2H3/i3+1,4+1,5+1,6+1

InChI key

VHILMKFSCRWWIJ-PQVJJBODSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service