Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

601373

Sigma-Aldrich

Rubidium-85Rb chloride

85 atom % (85Rb)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
85RbCl
CAS Number:
Molecular Weight:
120.36
UNSPSC Code:
12142200

isotopic purity

85 atom % (85Rb)

mass shift

M-1

SMILES string

[85Rb]Cl

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service