Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

597678

Sigma-Aldrich

3-Butenylamine hydrochloride

97%

Synonym(s):

1-Amino-3-butene hydrochloride, 3-Buten-1-amine hydrochloride, 4-Amino-1-butene hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
107.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

176-180 °C (lit.)

functional group

allyl
amine

SMILES string

Cl.NCCC=C

InChI

1S/C4H9N.ClH/c1-2-3-4-5;/h2H,1,3-5H2;1H

InChI key

MKEXLMVYRUNCQJ-UHFFFAOYSA-N

Application

3-Butenylamine hydrochloride can be used to synthesize 2-(3-butenylamino)-3-chloronaphthalene-1,4-dione via reaction with 2,3-dichloro-1,4-naphthoquinone in the presence of sodium bicarbonate.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of eight-, nine-, and ten-membered, nitrogen-containing quinone-fused heterocycles"
<BIG>Yerushalmi S, et al.</BIG>
Synthesis, 2007(02), 239- 242 (2007)
Caroline Mille et al.
Biosensors, 2(4), 433-447 (2012-01-01)
This work illustrates the enhancement of the sensitivity of the ELISA titration for recombinant human and native prion proteins, while reducing other non-specific adsorptions that could increase the background signal and lead to a low sensitivity and false positives. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service