Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

597058

Sigma-Aldrich

Ethyl nicotinate-13C6

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C2H9NO2
CAS Number:
Molecular Weight:
157.12
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

223-224 °C (lit.)

mp

8-10 °C

mass shift

M+6

SMILES string

[13CH3]COC(=O)[13c]1[13cH][13cH][13cH]n[13cH]1

InChI

1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3/i1+1,3+1,4+1,5+1,6+1,7+1

InChI key

XBLVHTDFJBKJLG-FJYIEFNNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service