Skip to Content
MilliporeSigma
All Photos(1)

Documents

596353

Sigma-Aldrich

Triethoxy(1-phenylethenyl)silane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiC(CH2)C6H5
CAS Number:
Molecular Weight:
266.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.4800 (lit.)

bp

264-265 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)C(=C)c1ccccc1

InChI

1S/C14H22O3Si/c1-5-15-18(16-6-2,17-7-3)13(4)14-11-9-8-10-12-14/h8-12H,4-7H2,1-3H3

InChI key

DKFGYGJKRWRIPR-UHFFFAOYSA-N

Application

A silicon-based nucleophile shown to be reactive in Pd-catalyzed cross-coupling reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.1 °F - closed cup

Flash Point(C)

90.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service