Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

596256

Sigma-Aldrich

trans-(2-Cyclohexylvinyl)boronic acid

95%

Synonym(s):

(E)-(2-Cyclohexylvinyl)boronic acid, trans-(2-Cyclohexylethenyl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BO2
CAS Number:
Molecular Weight:
154.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

Assay

95%

mp

106-111 °C (lit.)

storage temp.

2-8°C

SMILES string

OB(O)\C=C\C1CCCCC1

InChI

1S/C8H15BO2/c10-9(11)7-6-8-4-2-1-3-5-8/h6-8,10-11H,1-5H2/b7-6+

InChI key

FBRJOMMIILHLCG-VOTSOKGWSA-N

Application

Reactant for:
  • Pd-catalyzed coupling reactions
  • Chiral palladacycle-catalyzed asymmetric ring-opening reaction
  • Asymmetric Suzuki-Miyaura coupling

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service