Skip to Content
MilliporeSigma

595667

Sigma-Aldrich

α-Cyano-4-hydroxycinnamic acid-d7

97 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DOC6D4CD=C(CN)CO2D
CAS Number:
Molecular Weight:
196.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:


Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

97 atom % D

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

250-253 °C (dec.) (lit.)

mass shift

M+7

SMILES string

[2H]OC(=O)\C(C#N)=C(/[2H])c1c([2H])c([2H])c(O[2H])c([2H])c1[2H]

InChI

1S/C10H7NO3/c11-6-8(10(13)14)5-7-1-3-9(12)4-2-7/h1-5,12H,(H,13,14)/b8-5+/i1D,2D,3D,4D,5D/hD2

InChI key

AFVLVVWMAFSXCK-BMKBMMDNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service