Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

595349

Sigma-Aldrich

7-Methoxy-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

99-102 °C

functional group

ketone

SMILES string

COc1cccc2CCC(=O)c12

InChI

1S/C10H10O2/c1-12-9-4-2-3-7-5-6-8(11)10(7)9/h2-4H,5-6H2,1H3

InChI key

CZXBVBATQPHSSL-UHFFFAOYSA-N

General description

7-Methoxy-1-indanone can be synthesized by using chroman-4-one as the starting material.[1]

Application

7-Methoxy-1-indanone can be used to synthesize 10,15-Dihydro-4,9,14-trimethoxy-5H-diindeno[1,2-a;1′,2′-c]fluorene via acid-catalyzed trimerization.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Ring-substituted 11-oxo-11H-indeno [1, 2-b] quinoline-6-carboxamides with similar patterns of cytotoxicity to the dual topo I/II inhibitor DACA"
Deady.WL, et al.
Bioorganic & Medicinal Chemistry, 7(12), 2801- 2809 (1999)
"Synthesis of new C3h and C3v Truxene derivatives"
Gomez-Lor B, et al.
European Journal of Organic Chemistry, 2001(11), 2107-2114 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service